To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:T cell receptor beta chain
- Clonality:Monoclonal
- Clone:H57-597
- Host:Hamster
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:Tcrb
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Size:0.025 mg
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Tcrb
- Storage buffer:Phosphate-buffered aqueous solution, ≤0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Cat. no.:10766-044
- Supplier No.:77-216
Specifications
About this item
The H57-597 monoclonal antibody is specific for the beta chain of the mouse T cell Receptor (TCR). The crosslinking induces activation and proliferation of T cells, as a plate-bound or a soluble H57-597, or the plate-bound antibody can induce apoptosis, based on assay conditions. The beta chain of the TCR can combine with the alpha chain of the receptor to produce the alpha-beta TCR, which is expressed by the NKT cells, by the NK1.1+ thymocytes and most of the T cells. The beta chain does not react with the gamma-delta TCR-bearing cells, expressed by a small number of T cells. This antibody can be used as a phenotypic marker for the TCR beta expressing cells or for the functional purpose of TCR-mediated cell activation or apoptosis.
Type: Primary
Antigen: Tcrb
Clonality: Monoclonal
Clone: H57-597
Conjugation: APC
Epitope:
Host: Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse