To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:lymphocyte antigen 76
- Clonality:Monoclonal
- Clone:TER-119
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:Ly76
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:0.1 mg
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Ly76
- Storage buffer:Phosphate-buffered aqueous solution, ≤0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Tested applications:Functional
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Cat. no.:10765-410
- Supplier No.:77-071
Specifications
About this item
The TER-119 monoclonal antibody specifically binds to the mouse 52 kDa Glycophorin A expressed on the erythroid lineage cells, from the early proerythroblast to mature erythrocyte stages, in the fetal liver, adult bone marrow, newborn liver, yolk sac, and adult lymphoid organs. The TER-119 antibody is used, together with other antibodies, as a marker for hematopoietic lineages cells.
Type: Primary
Antigen: Ly76
Clonality: Monoclonal
Clone: TER-119
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: Rat IgG2b, kappa
Reactivity: Mouse