To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD8a molecule
- Clonality:Monoclonal
- Clone:OKT8
- Gene ID:P01732
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Antigen symbol:CD8A
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:25 tests
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:CD8A
- Storage buffer:Phosphate-buffered aqueous solution, ≤0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Cat. no.:10766-494
- Supplier No.:76-663
Specifications
About this item
The OKT8 monoclonal antibody specifically reacts with human CD8alpha molecule, a type I transmembrane glycoprotein of 32-34 kDa. CD8 alpha is a member of the Ig superfamily, expressed as a homodimer (CD8 alpha alpha) or as a heterodimer (CD8 alpha beta). CD8+ alpha beta T lymphocytes express both CD8 alpha alpha and CD8 alpha beta, while some T lymphocytes and the natural killer cells express only the homodimers. CD8 binds to MHC class I and influences the development and the activation of T lymphocytes. OKT8, RPA-T8, and HIT8a antibodies do not compete with each other for binding to peripheral leukocytes, meaning that that they do not recognize the same epitoPEor block each other by steric hindrance.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: OKT8
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: MouseIgG2a
Reactivity: Human