To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12)
- Clonality:Monoclonal
- Clone:TS2/16
- Gene ID:P05556
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:ITGB1
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 tests
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:ITGB1
- Storage buffer:Phosphate-buffered aqueous solution, ≤0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Cat. no.:10764-852
- Supplier No.:76-792
Specifications
About this item
The TS2/16 moncolonal antibody specifically reacts with human CD29 (Integrin beta1), a 130 kDA type I glycoprotein expressed on hematopoietic and non-hematopoietic cells. It forms the VLA-(1-6) molecules with integrin alpha-(1-6) and is involved in the adhesion between cell-cell and cell-matrix. The TS2/16 antibody has been found to activate beta 1 integrins.
Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: TS2/16
Conjugation: PE
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human