To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:selectin L
- Clonality:Monoclonal
- Clone:DREG-56
- Gene ID:P14151
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:SELL
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Size:0.05 mg
- Cross adsorption:No
- Format:purified antibody
- Form:liquid
- Antigen synonyms:SELL
- Storage buffer:Phosphate-buffered aqueous solution, ph7.2.
- Storage temperature:The product should be stored undiluted at 4 °C . Do not freeze.
- Shipping temperature:4 °C
- Tested applications:Functional
- Purification:The monoclonal antibody was purified utilizing affinity chromatography. The endotoxin level is determined by LAL test to be less than 0.01 EU/µg of the protein.
- Cat. no.:10765-336
- Supplier No.:76-211
Specifications
About this item
The DREG-56 monoclonal antibody specifically reacts with human CD62L, also known as L-selectin and LECAM-1. It mediates the migration of lymphocytes to the site of inflammation and their return to the peripheral lymphoid tissues and to the HEV (high endothelial venules). CD62L is expressed on monocytes, NK cells, neutrophils, granulocytes, and subsets of T and B cells. It is reported that the DREG-56 antibody inhibits the binding of lymphocytes to HEV tissue in frozen peripheral tissue.
Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: DREG-56
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human