To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:histone deacetylase 6
- Clonality:Polyclonal
- Gene ID:Q9UBN7
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:HDAC6
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.02 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:HDAC6
- Storage buffer:HDAC6 antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 55 kDa Observed: 32 kDa
- Storage temperature:HDAC6 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:HDAC6 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human HDAC6. The immunogen is located within the last 50 amino acids of HDAC6.
- Purification:HDAC6 antibody is affinity chromatography purified via peptide column.
- Cat. no.:10752-196
- Supplier No.:7901_S
Specifications
About this item
The histone deacetylase (HDAC) family contains multiple members which are divided into four classes. Class II of the HDAC family comprises six members, HDAC4, 5, 6, 7, 9 and 10, each of which appear to have tissue-specific roles. HDAC6 contains an internal duplication of two catalytic domains which appear to function independently of each other. HDAC6 has been shown to be part of the microtubule network and acts as a specific alpha-tubulin deacetylase, and has been suggested to be a potential therapeutic target in neurodegenerative disease.
Type: Primary
Antigen: HDAC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse