To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:D4, zinc and double PHD fingers, family 3
- Clonality:Polyclonal
- Gene ID:Q92784
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:DPF3
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.02 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:DPF3
- Storage buffer:DPF3 antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 39 kDa Observed: 50 kDa
- Storage temperature:DPF3 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:DPF3 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human DPF3. The immunogen is located within amino acids 220 - 270 of DPF3.
- Purification:DPF3 antibody is affinity chromatography purified via peptide column.
- Cat. no.:10752-090
- Supplier No.:7559_S
Specifications
About this item
The DPF3 protein, also known as Cerd4, is a member of the d4 gene family of transcription modulators that also includes DPF1/Neud4 and DPF2/Requiem. DPF3 has been shown to be a epigenetic key factor for heart and muscle development and can bind to methylated and acetylated lysine residues of histone 3 and 4, suggesting that DPF3 may play a role in recruiting chromatin remodeling complexes to acetylated histones. Two isoforms of DPF3, DPF3a and DPF3b, are required as transcriptional co-activators in SWI/SNF complex-dependent activation of the NF-kappaB RelA/p50 heterodimer.
Type: Primary
Antigen: DPF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse