To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:KN motif and ankyrin repeat domains 3
- Clonality:Polyclonal
- Gene ID:Q6NY19
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:KANK3
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.02 mg
- Western blot:Yes
- Epitope:near the N-Terminus
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:KANK3
- Storage buffer:KANK3 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 90 kDa Observed: 100 kDa
- Storage temperature:KANK3 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:KANK3 antibody was raised against an 18 amino acid peptide near the amino terminus of human KANK3 . The immunogen is located within amino acids 50 - 100 of KANK3.
- Purification:KANK3 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10751-952
- Supplier No.:7311_S
Specifications
About this item
KANK3 Antibody: Ankyrins are membrane adaptor molecules that play important roles in the control of cytoskeleton formation by regulating actin polymerization. Like other members of the KANK family, KANK3 (KN motif and ankyrin repeat domain-containing protein 3), is thought to play a role in the formation of actin stress fibers. In zebrafish, the homolog of KANK3 interacts with the adaptor protein Numb, a protein implicated in multiple basic cellular processes, and is essential for epidermal integrity and neurulation, suggesting that KANK3 may play a similar role in higher organisms.
Type: Primary
Antigen: KANK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse