To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:DEAH (Asp-Glu-Ala-His) box polypeptide 36
- Clonality:Polyclonal
- Gene ID:Q9H2U1
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:DHX36
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.02 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:DHX36
- Storage buffer:DHX36 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 111 kDa
- Storage temperature:DHX36 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:Rabbit polyclonal DHX36 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human DHX36. The immunogen is located within the last 50 amino acids of DHX36.
- Purification:DHX36 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10751-840
- Supplier No.:7163_S
Specifications
About this item
DHX36 Antibody: DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp, are putative RNA helicases implicated in several cellular processes involving modifications of RNA secondary structure. DHX36 (DEAH box protein 36), also known as MLE-like protein 1 and RNA helicase associated with AU-rich element ARE (RHAU), belongs to RNA helicase of the DEAH family and may function in sex development and spermatogenesis. It is expressed in testis and is evolutionary conserved with true orthologs in almost all animal species. DHX36 plays a role in degradation and deadenylation of mRNAs containing in their 3'-UTR the consensus ARE sequence element. DHX36 is required for early embryogenesis.
Type: Primary
Antigen: DHX36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse