To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:neurogenin 2
- Clonality:Polyclonal
- Gene ID:Q9H2A3
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NEUROG2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:0.02 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:NEUROG2
- Storage buffer:NGN2 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:30 kDa
- Storage temperature:NGN2 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:Rabbit polyclonal NGN2 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human NGN2. The immunogen is located within the last 50 amino acids of NGN2.
- Purification:NGN2 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10751-798
- Supplier No.:7091_S
Specifications
About this item
NGN2 Antibody: Neurogenin-2 (NGN2) is a neural-specific basic helix-loop-helix (bHLH) transcription factor that can specify a neuronal fate on ectodermal cells and is expressed in neural progenitor cells within the developing central and peripheral nervous systems. NGN2 is thought to work with Nurr1 to play a role in the differentiation and survival of midbrain dopaminergic neurons. It has also been suggested for use in human embryonic neural progenitors as a graft for spinal cord injuries
Type: Primary
Antigen: NEUROG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat