To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:patched domain containing 2
- Clonality:Polyclonal
- Gene ID:Q9P2K9
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:PTCHD2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.02 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:PTCHD2
- Storage buffer:PTCHD2 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:PTCHD2 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:PTCHD2 antibody was raised against a 19 amino acid synthetic peptide from near the center of human PTCHD2. The immunogen is located within amino acids 780 - 830 of PTCHD2.
- Purification:PTCHD2 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10751-514
- Supplier No.:6637_S
Specifications
About this item
PTCHD2 Antibody: PTCHD2, also known as DISP3, is the third of three known homologs of the D. melanogaster protein Dispatched. It is a multi-transmembrane protein containing two PTCH/DISP domains and is thought to be involved in the release of lipid-anchored secreted proteins. Like DISP1 and DISP2, DISP3 has been implicated in signaling pathways during embryogenesis, tissue regeneration, and carcinogenesis. It is highly expressed in Purkinje cells, hippocampal neurons, and retinal ganglion cells. Recently, it has been shown that PTCHD2 localizes within the endoplasmic reticulum and colocalizes with cholesterol, and given that its expression is regulated by thyroid hormone (T3), it has been suggested that DISP3 may be a link between thyroid hormone and cholesterol metabolism.
Type: Primary
Antigen: PTCHD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat