To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:actin filament associated protein 1-like 1
- Clonality:Polyclonal
- Gene ID:Q8TED9
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:AFAP1L1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:0.02 mg
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:AFAP1L1
- Storage buffer:AFAP1L Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:AFAP1L antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:AFAP1L1 antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human AFAP1L1. The immunogen is located within amino acids 70 - 120 of AFAP1L.
- Tested applications:Immunocytochemistry
- Purification:AFAP1L Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10751-468
- Supplier No.:6559_S
Specifications
About this item
AFAP1L Antibody: The actin filament-associated protein 1 (AFAP1)-like 1 protein is one of three members in the AFAP1 family of adaptor proteins. Like AFAP1, AFAP1L1 is an actin binding protein that has a role in actin cytoskeleton arrangement, but AFAP1L1 is thought to have a unique function distinct from AFAP1. AFAP1L1 colocalizes with cortactin and localizes to invadosomes, highly dynamic, actin-rich adhesion structures harboring metalloproteases, suggesting that AFAP1L1 affects invadosome formation (1, 4). Recent reports suggest that AFAP1L1 can be used as a prognostic marker for spindle cell sarcomas.
Type: Primary
Antigen: AFAP1L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse