To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 35, member D3
- Clonality:Polyclonal
- Gene ID:Q5M8T2
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:SLC35D3
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:0.02 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:SLC35D3
- Storage buffer:SLC35D3 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 46 kDa Observed: 45 kDa
- Storage temperature:SLC35D3 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:SLC35D3 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human SLC35D3. The immunogen is located within the last 50 amino acids of SLC35D3.
- Tested applications:Immunocytochemistry
- Purification:SLC35D3 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10751-456
- Supplier No.:6547_S
Specifications
About this item
SLC35D3 Antibody: The solute carrier family SLC35 consists of at least 17 proteins that act as nucleotide sugar transporters localized to the Golgi apparatus and endoplasmic reticulum. The novel protein SLC35D3 is highly homologous to SLC35D1 and SLC35D2, both of which transport UDP-glucuronic acid and UDP-N-acetylgalactosamine, suggesting that SLC35D3 is also involved in the transport of nucleotide sugars. It has been suggested that SLC35D3 regulates platelet dense granules, lysosome-related organelles which contain high concentrations of several biologically important low molecular weight molecules necessary for normal blood homeostasis.
Type: Primary
Antigen: SLC35D3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse