To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Rho GTPase activating protein 39
- Clonality:Polyclonal
- Gene ID:Q9C0H5
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ARHGAP39
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.02 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:ARHGAP39
- Storage buffer:ARHGAP39 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:ARHGAP39 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:ARHGAP39 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human ARHGAP39. The immunogen is located within amino acids 880 - 930 of ARHGAP39.
- Purification:ARHGAP39 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10751-394
- Supplier No.:6445_S
Specifications
About this item
ARHGAP39 Antibody: Rho GTPases are important regulators of the actin cytoskeleton and consequently influence the shape and migration of cells. GTPase-activating proteins (GAPs) accelerate the intrinsic rate of GTP hydrolysis of Ras-related proteins. ARHGAP39 (Rho GTPase activating protein 39) is a 1, 083 amino acid nuclear protein that contains one MyTH4 domain, one Rho GAP domain and two WW domains. It is encoded by a gene located on human chromosome 8, which is associated with a variety of diseases and malignancies. ARHGAP39 regulates robo signaling and plays important roles in axon guidance.
Type: Primary
Antigen: ARHGAP39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat