To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:potassium channel, subfamily K, member 13
- Clonality:Polyclonal
- Gene ID:Q9HB14
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:KCNK13
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.02 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:KCNK13
- Storage buffer:KCNK13 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:KCNK13 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:KCNK13 antibody was raised against a 15 amino acid synthetic peptide near the center of human KCNK13. The immunogen is located within amino acids 150 - 200 of KCNK13.
- Purification:KCNK13 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10751-380
- Supplier No.:6423_S
Specifications
About this item
KCNK13 Antibody: The closely related proteins KCNK13 and KCNK12 (also known as THIK1 and 2) are the first two members of a novel two pore-forming P domains K+ channels subfamily. The pore loop domain, a highly conserved region common to all potassium channels, is involved in determining potassium ion selectivity. Members of this family are all characterized by four transmembrane domains and may function to help influence the resting membrane potential of cells. KCNK13 is expressed mainly in the brain, but is also observed in kidneys. KCNK13 has been suggested to be a candidate for the Cs+-permeable K+ channel activated by GABA(B) receptors.
Type: Primary
Antigen: KCNK13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat