To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:zinc finger, MIZ-type containing 1
- Clonality:Polyclonal
- Gene ID:Q9ULJ6
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:ZMIZ1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:0.02 mg
- Western blot:Yes
- Epitope:near the N-Terminus
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:ZMIZ1
- Storage buffer:ZIMP10 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:ZIMP10 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:ZIMP10 antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human ZIMP10. The immunogen is located within amino acids 90 - 140 of ZIMP10.
- Tested applications:Immunocytochemistry
- Purification:ZIMP10 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10751-274
- Supplier No.:6237_S
Specifications
About this item
ZIMP10 Antibody: ZIMP10, also known as ZMIZ1, is a novel PIAS (protein inhibitor of activated signal transducer and activator of transcription)-like protein initially identified as a transcriptional co-activator of the androgen receptor (AR). ZIMP10 and the related protein ZIMP7 interact with PIAS3 and enhances AR-mediated transcription. Later experiments showed that ZIMP10 is also a co-activator of the p53 tumor suppressor. Mice deficient in ZIMP10 result in embryonic lethality by E10.5; these embryos reveal severe defects in the reorganization of the yolk sac vascular plexus, indicating that ZIMP10 plays an important role in proper vascular development.
Type: Primary
Antigen: ZMIZ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human