To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:zinc finger E-box binding homeobox 1
- Clonality:Polyclonal
- Gene ID:P37275
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:ZEB1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.02 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:ZEB1
- Storage buffer:ZEB1 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 124 kDa Observed: 130 kDa
- Storage temperature:ZEB1 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:ZEB1 antibody was raised against a 15 amino acid synthetic peptide near the center of human ZEB1. The immunogen is located within amino acids 530 - 580 of ZEB1.
- Tested applications:Immunocytochemistry
- Purification:ZEB1 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10751-000
- Supplier No.:5825_S
Specifications
About this item
ZEB1 Antibody: ZEB1, initially identified as the delta-crystallin enhancer binding protein delta EF1, is a DNA-binding protein that binds to a modified E-box sequence and has been implicated in postgastrulation embryogenesis. ZEB1 binds to the promoter of several hemapoietic genes, including interleukin-2, CD4, GATA-3, and alpha-integrin, and mice in which ZEB1 has been targeted show thymic atrophy, and severe defects in lymphocyte differentiation. Recent evidence suggests that ZEB1 also regulates the accumulation of adipose tissue and may play a role in obesity. Mutations in this gene have been associated with late-onset Fuchs endothelial corneal dystrophy.
Type: Primary
Antigen: ZEB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human