To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:kelch-like 25 (Drosophila)
- Clonality:Polyclonal
- Gene ID:Q9H0H3
- Host:Chicken
- Isotype:IgY
- Reactivity:Human
- Antigen symbol:KLHL25
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:0.02 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:KLHL25
- Storage buffer:ENC-2 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:ENC-2 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:ENC-2 antibody was raised against a 14 amino acid synthetic peptide near the center of human ENC-2. The immunogen is located within amino acids 230 - 280 of ENC-2.
- Tested applications:Immunocytochemistry
- Purification:ENC-2 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10750-902
- Supplier No.:5695_S
Specifications
About this item
ENC-2 Antibody: The ectoderm-neural cortex-2 (ENC-2) protein a member of the BTB/PDZ Kelch protein family displaying significant homology to ENC-1, an actin-binding protein involved in neural fate specification during embryogenesis and adipocyte maturation. The exact function of ENC-2 has yet to be determined, but it has been suggested to be involved in the differentiation of neuronal precursor cells into neurons and astrocytes.
Type: Primary
Antigen: KLHL25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human