Order Entry
ContactUsLinkComponent
Anti-FAM212A Rabbit Polyclonal Antibody
Anti-FAM212A Rabbit Polyclonal Antibody
Catalog #: 10750-756
Supplier:  Prosci
FAM212A antibody
Anti-FAM212A Rabbit Polyclonal Antibody
Catalog #: 10750-756
Supplier:  Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Family with sequence similarity 212 member A
  • Clonality:
    Polyclonal
  • Gene ID:
    Q96EL1
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Antigen symbol:
    FAM212A
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Size:
    0.02 mg
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Antigen synonyms:
    FAM212A
  • Storage buffer:
    FAM212A antibody is supplied in PBS containing 0.02% sodium azide.
  • Molecular weight:
    Predicted: 32 kDa Observed: 33 kDa
  • Storage temperature:
    FAM212A antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
  • Concentration:
    1 mg/mL
  • Shipping temperature:
    4 °C
  • Immunogen:
    FAM212A antibody was raised against a 15 amino acid peptide near the carboxy terminus of human FAM212A. The immunogen is located within the last 50 amino acids of FAM212A.
  • Purification:
    FAM212A antibody is affinity chromatography purified via peptide column.
  • Cat. no.:
    10750-756
  • Supplier No.:
    5531_S

Specifications

About this item

FAM212A, initially identified as INCA (induced in neural crest by AP-2alpha) in xenopus, is expressed primarily in neural crest cells and their derivatives. FAM212A can associate with the P21-kinase protein PAK4 and this association modulates cytoskeletal dynamics. In mammals, FAM212A is thought to play a role in neural tube closure; a significant fraction of FAM212A-null mice exhibited exencephaly.

Type: Primary
Antigen: FAM212A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Customers Who Bought This Also Bought