To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:ankyrin repeat domain 27 (VPS9 domain)
- Clonality:Polyclonal
- Gene ID:Q96NW4
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:ANKRD27
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.02 mg
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:ANKRD27
- Storage buffer:VARP Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:VARP antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:VARP antibody was raised against a 15 amino acid synthetic peptide near the center of human VARP. The immunogen is located within amino acids 640 - 690 of VARP.
- Purification:VARP Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10750-696
- Supplier No.:5451_S
Specifications
About this item
VARP Antibody: The VPS9 ankyrin repeat protein (VARP) binds to the Rab21, a guanine nucleotide exchange factor that plays an essential role in endocytic trafficking. VARP localizes to early endosomes and is thought to regulate endosome dynamics. VARP also interacts with TI-VAMP/VAMP7, a vesicular SNARE that mediates an exocytic pathway that is crucial to neurite growth. Depletion of VARP by RNA interference impairs neurite growth, suggesting that VARP is a positive regulator of neurite growth.
Type: Primary
Antigen: ANKRD27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human