To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:lacZ beta-D-galactosidase
- Clonality:Polyclonal
- Gene ID:Q8FKG6
- Host:Chicken
- Isotype:IgY
- Reactivity:Bacteria
- Antigen symbol:lacZ
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.02 mg
- Western blot:Yes
- Epitope:near the N-Terminus
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:lacZ
- Storage buffer:beta-Galactosidase Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:Antibody can be stored at 4 °C for three months and -20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:beta-gal antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of <i>E. coli</i> beta-gal. The immunogen is located within amino acids 20 - 70 of beta-Galactosidase.
- Purification:beta-Galactosidase Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10750-510
- Supplier No.:5155_S
Specifications
About this item
beta-Galactosidase Antibody: Beta-galactosidase (beta-gal), an inducible enzyme that catalyses the hydrolysis of lactose and other beta-galactosides into monosaccharides, is a product of the LacZ operon, and is a homo-tetrameric protein consisting of four identical subunits of approximately 114 kDa each. The enzyme is commonly used as a reporter gene in molecular biology to assess the efficiency of transfection.
Type: Primary
Antigen: lacZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Bacteria