To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:caspase 4, apoptosis-related cysteine peptidase
- Clonality:Polyclonal
- Gene ID:O75601
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:CASP4
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.02 mg
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:CASP4
- Storage buffer:Caspase-13 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:43 kDa
- Storage temperature:Caspase-13 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:Caspase-13 antibody was raised against a 15 amino acid peptide near the amino terminus of bovine Caspase-13. The immunogen is located within amino acids 80 - 130 of Caspase-13.
- Purification:Caspase-13 Antibody is Ion exchange chromatography purified.
- Cat. no.:10749-364
- Supplier No.:2245_S
Specifications
About this item
Caspase-13 Antibody: Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death is finally caused by members of the caspase family of proteases and caspase activated DNases. A novel member in the caspase family was recently identified and designated ERICE (for Evolutionarily Related Interleukin-1 beta Converting Enzyme) and caspase-13. Caspase-13 belongs to the ICE subfamily of caspases. Overexpression of caspase-13 induces apoptosis. Caspase-13 was activated by caspase-8, which is a key enzyme in death receptor induced apoptosis. Caspase-13 is expressed in a variety of human tissues and cell lines.
Type: Primary
Antigen: CASP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat