To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:cell death-inducing DNA fragmentation factor, alpha subunit-like effector B
- Clonality:Polyclonal
- Gene ID:O70303
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:Cideb
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.02 mg
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:Cideb
- Storage buffer:CIDE-B Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:25 kDa
- Storage temperature:CIDE-B antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:CIDE-B antibody was raised against a 16 amino acid peptide near the carboxy terminus of murine CIDE-B. The immunogen is located within the last 50 amino acids of CIDE-B.
- Purification:CIDE-B Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10749-288
- Supplier No.:2091_S
Specifications
About this item
CIDE-B Antibody: Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family of proteases. These death signals finally cause the degradation of chromosomal DNA by activated DNase. DFF45/ICAD has been identified as inhibitor of caspase activated DNase DFF40/CAD. DFF45 related proteins CIDE-A and CIDE-B (for cell death-inducing DFF-like effector A and B) were recently identified. CIDE contains a new type of domain termed CIDE-N, which has high homology with the regulatory domains of DFF45/ICAD and DFF40/CAD. Expression of CIDE-B induces apoptosis, which is inhibited by DFF45. CIDE-B is a DFF45-inhibitable effector that promotes cell death and DNA fragmentation. CIDE-B is expressed mainly in liver and at lower levels in spleen, kidney, peripheral blood lymphocytes, and bone marrow.
Type: Primary
Antigen: Cideb
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse