To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:calcium channel, voltage-dependent, T type, alpha 1H subunit
- Clonality:Polyclonal
- Gene ID:O95180
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:CACNA1H
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.02 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:CACNA1H
- Storage buffer:CACNA1H antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 259 kDa Observed: 260 kDa
- Storage temperature:CACNA1H antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:CACNA1H antibody was raised against a 16 amino acid peptide near the center of human CACNA1H. The immunogen is located within amino acids 1010 - 1060 of CACNA1H.
- Purification:CACNA1H antibody is affinity chromatography purified via peptide column.
- Cat. no.:10749-012
- Supplier No.:8029_S
Specifications
About this item
The calcium channel protein CACNA1H is a T-type member of the alpha-1 subunit family that is part of the voltage-dependent calcium channel complex which mediates the influx of calcium ions into the cell upon membrane polarization. CACNA1H is a subunit of Cav3.2 T-type calcium channel that is involved in neurological disorders such as epilepsy and pain. CACNA1H associates with the caveolin protein Cav-3 and it is thought that Cav-3 regulates the Protein Kinase A (PKA) modulation of CACNA1H-containing Cav3.2 T-type calcium channels.
Type: Primary
Antigen: CACNA1H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat