Tryptic Soy Broth, MilliporeSigma
Catalog #: CA1.05459.9025
Supplier: MilliporeSigma
CAS Number:
Specifications
- Packaging:Fibre carton
- Description:Tryptic Soy Broth
- Size:25 kg
- Solubility:30 g/L (20 °C)
- Density:570 kg/m³
- Storage:+15 °C…+25 °C
- Cat. no.:CA1.05459.9025
- Supplier No.:1.05459.9025
Specifications
About this item
Casein-peptone soymeal-peptone broth for microbiology (According harm. EP/USP/JP and ISO). Pepton from casein 17.0; Pepton from soymeal 3.0; D(+)-Glucose monohydrate 2.5; Sodium chloride 5.0; di-Potassium hydrogen phosphate 2.5.
- Clear after seven-day stability test at room temperature
- 10 to 100 inoculum on reference medium Staphylococcus aureus ATCC 6538 (WDCM 00032)
- 10 to 100 inoculum on reference medium Bacillus subtilis ATCC 6633 (WDCM 00003)
- 10 to 100 inoculum on reference medium Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)
- 10 to 100 inoculum on reference medium Escherichia coli ATCC 8739 (WDCM 00012)
- 10 to 100 inoculum on reference medium Streptococcus pneumoniae ATCC 6301
- 10 to 100 inoculum on reference medium Salmonella typhimurium ATCC 14028 (WDCM 00031)
- 10 to 100 inoculum on reference medium Staphylococcus aureus ATCC 25923 (WDCM 00034)
- 10 to 100 inoculum on reference medium Staphylococcus epidermidis ATCC 12228 (WDCM 00036)
- 10 to 100 inoculum on reference medium Candida albicans ATCC 2091 (WDCM 00055)
- 10 to 100 inoculum on reference medium Candida albicans ATCC 10231 (WDCM 00054)
- 10 to 100 inoculum on reference medium Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053)
- Visible growth in 18 to 24 hours at 30 to 35°C in Staphylococcus aureus ATCC 6538 (WDCM 00032)
- Visible growth in 18 to 24 hours at 30 to 35°C in Bacillus subtilis ATCC 6633 (WDCM 00003)
- Visible growth in 18 to 24 hours at 30 to 35°C in Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)
- Visible growth in 18 to 24 hours at 30 to 35°C in Escherichia coli ATCC 8739 (WDCM 00012)
- Visible growth in 18 to 24 hours at 30 to 35°C in Streptococcus pneumoniae ATCC 6301
- Visible growth in 18 to 24 hours at 30 to 35°C in Salmonella typhimurium ATCC 14028 (WDCM 00031)
- Visible growth in 3 days at 20 to 25°C in Staphylococcus aureus ATCC 6538 (WDCM 00032)
- Visible growth in 3 days at 20 to 25°C in Staphylococcus aureus ATCC 25923 (WDCM 00034)
- Visible growth in 3 days at 20 to 25°C in Staphylococcus epidermidis ATCC 12228 (WDCM 00036)
- Visible growth in 3 days at 20 to 25°C in Bacillus subtilis ATCC 6633 (WDCM 00003)
- Visible growth in 5 days at 20 to 25°C in Candida albicans ATCC 2091 (WDCM 00055)
- Visible growth in 5 days at 20 to 25°C in Candida albicans ATCC 10231 (WDCM 00054)
- Visible growth in 5 days at 20 to 25°C in Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053)
Customers Who Bought This Also Bought