Order Entry
ContactUsLinkComponent
SABOURAUD 4% Dextrose Agar, MilliporeSigma
SABOURAUD 4% Dextrose Agar, MilliporeSigma
Catalog #: CA1.05438.5000
Supplier:  MilliporeSigma
CAS Number:  
SABOURAUD 4% Dextrose Agar, MilliporeSigma
Catalog #: CA1.05438.5000
Supplier:  MilliporeSigma
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Packaging:
    Plastic Drum
  • Description:
    SABOURAUD 4% Dextrose Agar, EMD Millipore
  • Size:
    5 kg
  • Solubility:
    65 g/L (100 °C)
  • Density:
    680 kg/m³
  • Incubation:
    7 days; 28°C
  • Storage:
    Store at +15°C to +25°C
  • pH:
    5.6 (65 g/L, H₂O, 25 °C) (after autoclaving)
  • Cat. no.:
    CA1.05438.5000
  • Supplier No.:
    1.05438.5000

Specifications

About this item

In North America 1.05438.5000 replaces and is identical to 1.05438.5007

  • Clear, yellowish-brown appearance
  • Fair to very good growth in Trichophyton mentagrophytes ATCC 18748
  • Fair to good growth in Trichophyton rubrum ATCC 28188
  • Fair to good growth in Trichophyton ajelloi ATCC 28454
  • Fair to very good growth in Microsporum gallinae ATCC 12108
  • Good to very good growth in Microsporum canis ATCC 36299
  • Good to very good growth in Geotrichum candidum DSM 1240
  • Good to very good growth in Penicillium commune ATCC 10428
Customers Who Bought This Also Bought