ContactUsLinkComponent
VWR® Freezer Box for Centrifuge Tubes
VWR® Freezer Box for Centrifuge Tubes
Catalog #: 10002-734
Supplier:  VWR International
Freezer Box
VWR® Freezer Box for Centrifuge Tubes
Catalog #: 10002-734
Supplier:  VWR International
Specifications
  • Color:
    Clear, Blue Base
  • Description:
    VWR® Freezer Box for Centrifuge Tubes
  • Cat. no.:
    10002-734
Specifications
About this item
VWR® Freezer Box for Centrifuge Tubes features polycarbonate construction and is supplied with lid.
  • Suitable over a range of temperatures
  • Polycarbonate construction
The box is suitable for use in temperatures as low as -190°C (-310°F). Each box will accommodate 25 x 5 mL centrifuge tubes.
Customers Who Bought This Also Bought