OmniTrace Ultra™ Dispenser, MilliporeSigma
Supplier: MilliporeSigma
Certificates
About this item
For easy, safe and reliable dispensing of OmniTrace Ultra™ high purity acids without contamination
All wetted parts are constructed of fluoropolymer resin. Certificate of Performance included with each dispenser.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.