Sand Quartz Slide
Catalog #: 470177-662
Supplier: Avantor
Specifications
- Description:Sand Quartz
- Cat. no.:470177-662
- Supplier No.:WARD470177-662
- Orientation:Whole mount
Specifications
About this item
Observe the Main Constituent of Sand
- Identifying features clearly distinguishable
- Rigorous quality control standards
Quartz is a brilliant, hexagonally crystalline mineral, silicon dioxide (SiO2).
Customers Who Bought This Also Bought