Order Entry
ContactUsLinkComponent
Human Recombinant Siglec6 (from HEK293 Cells)
Human Recombinant Siglec6 (from HEK293 Cells)
Catalog #: 10798-368
Supplier:  Prosci
CAS Number:  
SIGLEC6 Recombinant Protein
Human Recombinant Siglec6 (from HEK293 Cells)
Catalog #: 10798-368
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.1 mg
  • Tag sequence:
    C-Fc Tag
  • Biological activity:
    Measured by its ability to bind biotinylated Neu5Ac alpha 2-6GalNAc alpha (sialyl-Tn).
  • Protein synonyms:
    CDw327|CD33L|SIGLEC6|CD327|OBBP1|CD33L1|OB-BP1
  • Protein/peptide name:
    Siglec6
  • Purity:
    >92% as determined by SDS-PAGE.
  • Molecular weight:
    60.3 kDa
  • Formulation:
    Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
  • Tested applications:
    Western Blot
  • Cat. no.:
    10798-368
  • Supplier No.:
    96-692

Specifications

About this item

Sialic acid-binding Ig-like lectin 6 (SIGLEC6) is also known as CD antigen CD327, CD33 antigen-like 1 (CD33L or CD33L1), Obesity-binding protein 1 (OB-BP1). SIGLEC6 belongs to the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family, which contains two Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. SIGLEC6 mediates sialic-acid dependent binding to cells. SIGLEC6 binds to alpha-2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface.

Customers Who Bought This Also Bought