Order Entry
ContactUsLinkComponent
Human Recombinant Nectin 1 (from HEK293 Cells)
Human Recombinant Nectin 1 (from HEK293 Cells)
Catalog #: 10797-852
Supplier:  Prosci
CAS Number:  
Nectin 1 Recombinant Protein
Human Recombinant Nectin 1 (from HEK293 Cells)
Catalog #: 10797-852
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.1 mg
  • Tag sequence:
    His Tag
  • Protein synonyms:
    PVRL1|CD111|OFC7|PVRR|PVRR1|PRR|HVEC|ED4|CLPED1|HIgR|Nectin-1|SK-12|PRR1
  • Protein/peptide name:
    Nectin 1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    34.8 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot
  • Cat. no.:
    10797-852
  • Supplier No.:
    96-642

Specifications

About this item

Poliovirus receptor-related protein 1 (PVRL1) is also known as Herpes virus entry mediator C (HveC), Herpesvirus Ig-like receptor (HIgR), Nectin-1, CD antigen CD111, PVRL1 / Nectin-1 / CD111 belongs to the nectin family. PVRL1 / Nectin-1 is a membrane protein with three extracellular immunoglobulin domains, a single transmembrane helix and a cytoplasmic tail. The protein can mediate Ca2+-independent cellular adhesion further characterizing it as IgSF cell adhesion molecule (IgSF CAM). PVRL1 is adhesion molecule found in a wide range of tissues. PVRL1 / Nectin-1 promotes cell-cell contacts by forming homophilic or heterophilic trans-dimers. Heterophilic interactions have been detected between PVRL1/nectin-1 and PVRL3/nectin-3 and between PVRL1/nectin-1 and PVRL4/nectin-4.

Customers Who Bought This Also Bought