Order Entry
ContactUsLinkComponent
Human Recombinant ICAM1 (from HEK293 cells)
Human Recombinant ICAM1 (from HEK293 cells)
Catalog #: 10797-448
Supplier:  Prosci
CAS Number:  
ICAM 1 Recombinant Protein
Human Recombinant ICAM1 (from HEK293 cells)
Catalog #: 10797-448
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.2 mg
  • Tag sequence:
    C-Fc Tag
  • Protein synonyms:
    BB2|CD54|P3.58|ICAM1
  • Protein/peptide name:
    ICAM1
  • Purity:
    >98% as determined by SDS-PAGE.
  • Molecular weight:
    75.7 kDa
  • Formulation:
    Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
  • Tested applications:
    Western Blot
  • Cat. no.:
    10797-448
  • Supplier No.:
    96-400

Specifications

About this item

Inter-Cellular Adhesion Molecule 1 (ICAM-1) is also known as Cluster of Differentiation 54 (CD54), is a member of the immunoglobulin superfamily, and is a cell surface glycoprotein which is typically expressed in low concentrations on endothelial cells and cells of the immune system. The protein encoded by this gene is a type of intercellular adhesion molecule continuously present in low concentrations in the membranes of leukocytes and endothelial cells. Upon cytokine stimulation, the concentrations greatly increase. ICAM-1 can be induced by interleukin-1 (IL-1) and tumor necrosis factor alpha (TNF?) and is expressed by the vascular endothelium, macrophages, and lymphocytes. ICAM-1 is a ligand for LFA-1 (integrin), a receptor found on leukocytes. When activated, leukocytes bind to endothelial cells via ICAM-1/LFA-1 and then transmigrate into tissues. ICAM-1 has been implicated in subarachnoid hemorrhage (SAH). Levels of ICAM-1 are shown to be significantly elevated in patients with SAH over control subjects in many studies. ICAM-1 expressed by respiratory epithelial cells is also the binding site for rhinovirus, the causative agent of most common colds.

Customers Who Bought This Also Bought