Order Entry
ContactUsLinkComponent
HMGB1 Recombinant Protein, His Tag, ProSci
HMGB1 Recombinant Protein, His Tag, ProSci
Catalog #: 10797-434
Supplier:  Prosci
CAS Number:  
HMGB1 Recombinant Protein
HMGB1 Recombinant Protein, His Tag, ProSci
Catalog #: 10797-434
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.1 mg
  • Tag sequence:
    His Tag
  • Biological activity:
    Measured by its binding ability in a functional ELISA Immobilized human HMGB at 2μg/ml (100μl/well) can bind human AGER. The EC50 of human AGER is 0. 22μg/ml.
  • Protein synonyms:
    SBP-1|HMGB1|HMG3|HMG1
  • Protein/peptide name:
    Hmgb1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    25.7 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot
  • Cat. no.:
    10797-434
  • Supplier No.:
    96-393

Specifications

About this item

High-mobility group protein B1 (HMGB1) is also known as high-mobility group protein 1 (HMG-1) and amphoterin, is a member of the HMGB family consisting of three members, HMGB1, HMGB2 and HMGB3. HMGB1 is a non-histone architectural chromosomal protein ubiquitously present in all vertebrate nuclei and binds double-stranded DNA without sequence specificity. The mechanism of inflammation and damage is binding to TLR4, which mediates HMGB1-dependent activation of macrophage cytokine release. This positions HMGB1 at the intersection of sterile and infectious inflammatory responses. HMGB1 has been studied as a DNA vaccine adjuvant and a target for cancer therapy.

Customers Who Bought This Also Bought