Order Entry
ContactUsLinkComponent
Human Recombinant Cgp 39 (from HEK293 Cells)
Human Recombinant Cgp 39 (from HEK293 Cells)
Catalog #: 10797-204
Supplier:  Prosci
CAS Number:  
CGP 39 Recombinant Protein
Human Recombinant Cgp 39 (from HEK293 Cells)
Catalog #: 10797-204
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.1 mg
  • Tag sequence:
    His Tag
  • Biological activity:
    Measured by the ability of the immobilized protein to support the adhesion of FaDu human squamous cell carcinoma cells. When 50000 cells per well are added to rhCHI3-L1 coated plate, cell adhesion is enhanced in a dose dependent manner. The ED50 for this effect is typically 0. 8 – 3. 5 μg/mL.
  • Protein synonyms:
    CHI3L1|CGP-39|GP-39|YKL-40|hCGP-39|ASRT7
  • Protein/peptide name:
    Cgp 39
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    41.3 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot
  • Cat. no.:
    10797-204
  • Supplier No.:
    96-186

Specifications

About this item

Chitinase-3-like protein 1 (CHI3L1) is also known as Cartilage glycoprotein 39 (CGP-39), YKL-40, which belongs to the glycosyl hydrolase 18 family. CHI3L1 is present in activated macrophages, articular chondrocytes, synovial cells as well as in liver. CHI3L1 may play a role in defense against pathogens, or in tissue remodeling and also may play an important role in the capacity of cells to respond to and cope with changes in their environment.

Customers Who Bought This Also Bought