Order Entry
ContactUsLinkComponent
Human Recombinant FGF1 (from E. coli)
Human Recombinant FGF1 (from E. coli)
Catalog #: 10796-966
Supplier:  Prosci
CAS Number:  
FGF1 Recombinant Protein
Human Recombinant FGF1 (from E. coli)
Catalog #: 10796-966
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.2 mg
  • Tag sequence:
    Tag free
  • Biological activity:
    The bioactivity of rh-aFGF protein was determined in a BALB/c 3T3 mouse fibroblasts cell proliferation assay. The ED50 of each lot is between 50-200 pg/mL.
  • Protein synonyms:
    AFGF|ECGFA|ECGF-beta|FGF1|FGF-alpha|HBGF1|FGFA|GLIO703|ECGF|ECGFB
  • Protein/peptide name:
    FGF1
  • Purity:
    >98% as determined by SDS-PAGE.
  • Molecular weight:
    15.8 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot
  • Cat. no.:
    10796-966
  • Supplier No.:
    96-016

Specifications

About this item

Heparin-binding growth factor 1 is a protein that in humans is encoded by the FGF1 gene. The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF acidic is a potent growth factor for fibroblasts and endothelial cells. FGF acidic is involved in wound repair, angiogenesis, and development. FGF acidic is secreted from cells via an endoplasmic reticulum/Golgi independent mechanism. The ability of FGF acidic to bind to heparin sulfate is required for its ability to interact with FGF receptors and induce signaling. There are four distinct FGF receptors and each has multiple splice variants. FGF acidic binds with high affinity to many, but not all, FGFRs. Signaling cascades activated through FGF basic binding to FGFR include the ras-raf-MAPK, PLC?/PKC, and PI3K/Akt pathways.

Customers Who Bought This Also Bought