To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:2.5 U/µl
- Description:KOD DNA Polymerase
- Storage conditions:−20 °C
- Cat. no.:CA80511-382
- Supplier No.:71085-3
Specifications
About this item
KOD DNA Polymerase is a recombinant form of Thermococcus kodakaraensis KOD1 DNA polymerase.
- Higher fidelity, excellent for cloning
- Greater yield, its extension speed is fast
- Amplifies plasmid and lambda DNA upto 6 kbp and genomic DNA upto 2 kbp
- No truncated amplification products
It is a high fidelity thermostable DNA polymerase that amplifies target DNA up to 6 kbp with superior accuracy and yield for PCR applications. The enzyme's 3'→5' exonuclease-dependent proofreading activity results in a lower PCR mutation frequency than any other available DNA polymerase.
The elongation rate and processivity are 5 times and 10 to 15 times higher, respectively, than for Pfu DNA polymerase, resulting in highly accurate and robust yield, in a short reaction time. The enzyme generates blunt-ended PCR products suitable for cloning with various vector kits.