To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:ATG13 autophagy related 13 homolog
- Clonality:Polyclonal
- Gene ID:ATG13/9776; Atg13/51897
- Host:Rabbit
- Reactivity:Human,Mouse
- Antigen symbol:KIAA0652
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:400 µL
- Storage buffer:PBS with 0.09% Sodium azide
- Immunogen:Synthetic peptide from the N-terminal region of human KIAA0652 conjugated to KLH.
- Cat. no.:CAPIPA526923
- Supplier No.:PA526923
Specifications
About this item
This antibody is predicted to react with bovine based on sequence homology. Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein ATG1. ATG13 forms a complex with ULK1 and ULK2, the mammalian homologs of ATG1, and with FIP200. This complex is a target of TOR phosphorylation under normal conditions; inhibition of TOR by rapamycin or leucine deprivation leads to dephosphorylation of ATG13, ULK1 and ULK2, which then leads to autophagy. Knockdown of ATG13 inhibits autophagosome formation.
Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000
Type: Primary
Antigen: ATG13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse