To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:DEAD (Asp-Glu-Ala-Asp) box helicase 17
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:DDX17
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:400 µL
- Antigen synonyms:DEAD/H box 17|RH70|RP3-434P1.1|DEAD box protein 17|RNA-dependent helicase p72|P72|DEAD box protein p72|probable ATP-dependent RNA helicase DDX17|DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 17 (72kD)|DKFZp761H2016
- Storage buffer:PBS with 0.09% Sodium azide
- Immunogen:Synthetic peptide from the N-terminal region of human DDX17 conjugated to KLH.
- Cat. no.:CAPIPA526330
- Supplier No.:PA526330
Specifications
About this item
This antibody is predicted to react with mouse based on sequence homology. DDX17 box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and splicesosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This protein encodes a DEAD box protein, which is an ATPase activated by a variety of RNA species, but not by dsDNA. This protein, and that encoded by DDX5 gene, are more closely related to each other than to any other member of the DEAD box family.
Flow Cytometry: 1:10-1:50, Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000
Type: Primary
Antigen: DDX17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human