To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, Subfamily F, Member 1
- Clonality:Polyclonal
- Gene ID:ABCF1/23; Abcf1/224742
- Host:Rabbit
- Reactivity:Human,Mouse
- Antigen symbol:ABCF1
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:400 µL
- Antigen synonyms:ATP-binding cassette sub-family F member 1|ATP-binding cassette 50|ABC50|TNF-alpha-stimulated ABC protein|ABCF1|ABC27
- Storage buffer:PBS with 0.09% Sodium azide
- Immunogen:Synthetic peptide from the C-terminal region of human ABCF1 conjugated to KLH.
- Cat. no.:CAPIPA526286
- Supplier No.:PA526286
Specifications
About this item
This antibody is predicted to react with porcine based on sequence homology. The protein is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC proteins are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the GCN20 subfamily. Unlike other members of the superfamily, this protein lacks the transmembrane domains which are characteristic of most ABC transporters. This protein may be regulated by tumor necrosis factor-alpha and play a role in enhancement of protein synthesis and the inflammation process.
Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000
Type: Primary
Antigen: ABCF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human