To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Bromo Adjacent Homology Domain Containing 1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:BAHD1
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:400 µL
- Antigen synonyms:BAH Domain-Containing Protein 1|KIAA0945|Gm117|AL022997|AW541238
- Storage buffer:PBS with 0.09% Sodium azide
- Immunogen:Synthetic peptide from the C-terminal region of human BAHD1 conjugated to KLH.
- Cat. no.:CAPIPA526166
- Supplier No.:PA526166
Specifications
About this item
This antibody is predicted to react with mouse based on sequence homology. BAHD1 is a heterochromatin protein that acts as a transcription repressor and has the ability to promote the formation of large heterochromatic domains. It may act by recruiting heterochromatin proteins such as CBX5 (HP1 alpha), HDAC5 and MBD1. It represses IGF2 expression by binding to its CpG-rich P3 promoter and recruiting heterochromatin proteins.
Flow Cytometry: 1:10-1:50, Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000
Type: Primary
Antigen: BAHD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human