To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Olfactory Receptor, Family 2, Subfamily H Member 2
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:OR2H2
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:400 µL
- Storage buffer:PBS with 0.09% Sodium azide
- Immunogen:Synthetic peptide from the C-terminal region of human OR2H2 conjugated to KLH.
- Cat. no.:CAPIPA526044
- Supplier No.:PA526044
Specifications
About this item
OR2H2 is a odorant receptor.
Recommended Dilutions: Western Blot: 1:1000, Flow Cytometry: 1:10-1:50
Type: Primary
Antigen: OR2H2 (olfactory receptor, family 2, subfamily H, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human