To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Activating Transcription Factor 7
- Clonality:Polyclonal
- Gene ID:11016
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Bovine,Dog,Rat
- Antigen symbol:ATF7
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:ATFA
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-HVIMTPQSQSAGR
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89298-360
- Supplier No.:GTX89982
Specifications
About this item
Goat polyclonal antibody to ATF7
Approx 35kDa band seen in HeLa cell lysates. Recommended for use at 0.5-2µg/ml. Please note the band size is in agreement with previous experimental data (J Biol Chem 2001 Apr 27;276(17):13718-26). In paraffin embedded Human Testis shows staining of select spermatocytes in Human Testis. Recommended concentration, 3-5µg/ml. This antibody is expected to recognise all three reported isoforms (NP_001123531.1, NP_006847.1 and NP_001123532.1).
Type: Primary
Antigen: ATF7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Bovine, Dog, Rat