To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Intraflagellar Transport 74
- Clonality:Polyclonal
- Gene ID:80173
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat
- Antigen symbol:CMG1 / CCDC2 / IFT74 (intraflagellar transport 74 homolog (Chlamydomonas))
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:CMG1|CMG-1|CCDC2
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-KTIVDALHSTSGN
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89298-078
- Supplier No.:GTX89771
Specifications
About this item
Goat polyclonal antibody to CMG1 / CCDC2 / IFT74
Approx 70-75kDa band observed in lysates of cell line HEK293 (calculated MW of 69.2kDa according to AAK77221.1, but 51.4kDa according to NP_079379.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Momeni et al, BMC Neurol. 2006 Dec 13;6:44.; PMID: 17166276). Recommended concentration: 0.3-1µg/ml. In paraffin embedded Human Liver shows vesicular staining in cytoplasm of hepatocytes. Recommended concentration, 5-10µg/ml.
Type: Primary
Antigen: CMG1 / CCDC2 / IFT74 (intraflagellar transport 74 homolog (Chlamydomonas))
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: