To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Toll-interleukin 1 Receptor (TIR) Domain Containing Adaptor Protein
- Clonality:Polyclonal
- Gene ID:114609
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:TIRAP
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:TIRAP|OTTHUMP00000179130|toll-interleukin 1 receptor (TIR) domain-containing adapter protein|MyD88 adapter-like protein|adapter protein wyatt|Toll-like receptor adaptor protein|FLJ42305|Toll-interleukin 1 receptor domain-containing adaptor protein|Mal|wyatt
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-EGEGERDSATVSDL
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89297-962
- Supplier No.:GTX89676
Specifications
About this item
Goat polyclonal antibody to TIRAP / Mal (Isoform b)
Approx 30kDa band observed in Human Liver lysates (calculated MW of 25.3kDa according to NP_683708.1). Recommended concentration: 1-3µg/ml. This protein is expected to recognise isoform b (NP_683708.1) only.
Type: Primary
Antigen: TIRAP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human