To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:pan ADH (alcohol dehydrogenase 1A (class I) alpha Polypeptide)
- Clonality:Polyclonal
- Gene ID:124
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ADH1A
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:ADH1
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:STAGKVMKCKA
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89297-874
- Supplier No.:GTX89624
Specifications
About this item
Goat polyclonal antibody to pan ADH
Approx 38kDa band observed in Human Liver lysates (calculated MW of 39.9kDa according to NP_000658.1, NP_000659.2 and 000660.1). Recommended concentration: 1-3µg/ml. Customer found this product to work in IHC on Human Liver. This antibody is expected to recognise the alpha (ADH1A, NP_ 000658.1), the beta (ADH1B, NP_000659.2) and gamma (ADH1C, NP_000660.1) polypeptide variants of human alcohol dehydrogenase.
Type: Primary
Antigen: ADH1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human