To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:SECIS binding Protein 2
- Clonality:Polyclonal
- Gene ID:79048
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Dog
- Antigen symbol:SBP2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:SECISBP2|selenocysteine insertion sequence binding protein 2|RP11-89K14.1|OTTHUMP00000064929|OTTHUMP00000064930|OTTHUMP00000064931|OTTHUMP00000064932|SBP2 |DKFZp686C09169|SECIS binding protein 2
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-KERQERKQRLQEN
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89297-394
- Supplier No.:GTX89253
Specifications
About this item
Goat polyclonal antibody to SBP2
Approx 120kDa band detected in HFF cell lysates. Data kindly provided by Laura Papp and Kum Kum Khanna. The observed molecular weight corresponds to earlier findings in literature with different antibodies (Papp et al, Mol Cell Biol. 2006 Jul;26(13):4895-910; PMID: 16782878). Recommended concentration: 0.1-0.3µg/ml. In paraffin embedded Human Testis shows granular cytoplasm staining in Leydig cells. Recommended concentration, 3µg/ml.
Type: Primary
Antigen: SBP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Dog