To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Fibroblast Growth Factor 23
- Clonality:Polyclonal
- Gene ID:8074
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:FGF23
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:PHPTC|ADHR|HPDR2|FGFN|HYPF
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-RHTRSAEDDSERD
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89297-320
- Supplier No.:GTX89195
Specifications
About this item
Goat polyclonal antibody to FGF23
Approx 26-27kDa band observed in human brain (Hippocampus) lysates (calculated MW of 27.9kDa according to NP_065689.1). Recommended concentration: 0.5-1.5µg/ml. An additional band of unknown identity was also consistently observed at 37kDa. This band was successfully blocked by incubation with the immunising peptide. We would appreciate any feedback from people in the field - have any such results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported?
Type: Primary
Antigen: FGF23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human