To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Spleen focus forming virus (SFFV) proviral integration oncogene spi1
- Clonality:Polyclonal
- Gene ID:6688
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat
- Antigen symbol:SPI1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:spleen focus forming virus (SFFV) proviral integration oncogene spi1 |SPI-A|SPI1|hematopoietic transcription factor PU.1|SPI-1|PU.1|SPI-1 proto-oncogene|31 kDa transforming protein|SFPI1|hCG_25181
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-DLYQRQTHEYY
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89296-624
- Supplier No.:GTX88620
Specifications
About this item
Goat polyclonal antibody to PU.1/Spi1
Approx 40kDa band observed in lysates of DAUDI and K562 (calculated MW of 31.2kDa according to NP_001074016.1 and of 31.1kDa according to NP_003111.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Harendza S et al, J Biol Chem. 2000 Jun 30;275(26):19552-9.; PMID: 10867017). Recommended concentration: 0.03-0.1µg/ml. This antibody is expected to recognise both reported isoforms (NP_001074016.1; NP_003111.2).
Type: Primary
Antigen: SPI1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat