To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Deah (Asp-Glu-Ala-His) Box Helicase 9
- Clonality:Polyclonal
- Gene ID:1660
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Bovine,Rat,Mouse
- Antigen symbol:DHX9 / RHA (DEAH (Asp-Glu-Ala-His) box Polypeptide 9)
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:RHA|NDHII|DDX9|NDH2|LKP
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:TEGRNALIHKSSVNC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89296-392
- Supplier No.:GTX88436
Specifications
About this item
Goat polyclonal antibody to DHX9 / RHA
Approx. 140kDa band observed in nuclear lysates of cell line HeLa (calculated MW of 141kDa according to NP_001348.2). Recommended concentration: 1-3µg/ml.
Type: Primary
Antigen: DHX9 / RHA (DEAH (Asp-Glu-Ala-His) box Polypeptide 9)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Bovine, Rat