To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Nk2 Homeobox 5
- Clonality:Polyclonal
- Gene ID:1482
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Antigen symbol:CSX1 / NKX2-5 (NK2 transcription Factor related locus 5 (Drosophila))
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:VSD3|NKX4-1|NKX2.5|CHNG5|CSX1|NKX2E|CSX|HLHS2
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-PRAYSDPDPAKDPR
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89296-340
- Supplier No.:GTX88402
Specifications
About this item
Goat polyclonal antibody to CSX1 / NKX2-5
Approx 45kDa band observed in Human Heart lysates (calculated MW of 34.9kDa according to NP_004378.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Shiojima et al, Circ Res. 1996 Nov;79(5):920-9.; PMID: 8888684). Recommended concentration: 0.3-1µg/ml. This antibody is expected to recognize all three reported isoforms (NP_004378.1; NP_001159647.1; NP_001159648.1).
Type: Primary
Antigen: CSX1 / NKX2-5 (NK2 transcription Factor related locus 5 (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: